Assay Details
| Gene Symbol: postnb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203421.1 | NP_981966.1 | 20 - 21 | 2340 | 107 | |
| XM_009291791.3 | XP_009290066.1 | 22 - 23 | 2592 | 107 | ||
| XM_021481350.1 | XP_021337025.1 | 21 - 22 | 2547 | 107 | ||
| GenBank mRNA | AB126325.1 | - | 20 - 21 | 2340 | 107 | |
| BC162274.1 | - | 20 - 21 | 2394 | 107 | ||
More Information
Set Membership: |
|

postnb
Genomic Map