Assay Details
| Gene Symbol: hsd17b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_205584.2 | NP_991147.2 | 3 - 4 | 497 | 61 | |
| XM_021473358.1 | XP_021329033.1 | 3 - 4 | 644 | 61 | ||
| GenBank mRNA | AY306005.1 | - | 3 - 4 | 497 | 61 | |
| BC163707.1 | - | 3 - 4 | 477 | 61 | ||
| BC163709.1 | - | 3 - 4 | 477 | 61 | ||
More Information
Set Membership: |
|

hsd17b1
Genomic Map