Assay Details
Gene Symbol: znf410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205653.1 | NP_991216.1 | 6 - 7 | 658 | 104 | |
XM_005170226.4 | XP_005170283.1 | 6 - 7 | 781 | 104 | ||
XM_005170227.4 | XP_005170284.1 | 6 - 7 | 781 | 104 | ||
XM_005170228.3 | XP_005170285.1 | 6 - 7 | 781 | 104 | ||
XM_009295041.3 | XP_009293316.1 | 6 - 7 | 781 | 104 | ||
XM_017352450.2 | XP_017207939.1 | 6 - 7 | 781 | 104 | ||
GenBank mRNA | BC065642.1 | - | 6 - 7 | 658 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
