Assay Details
Gene Symbol: pdpk1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_205699.1 | 12 - 13 | 1399 | 81 | ||
GenBank mRNA | BC065959.1 | - | 12 - 13 | 1399 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
