Assay Details
| Gene Symbol: rab18a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_206830.1 | NP_996661.1 | 6 - 7 | 539 | 65 | |
| GenBank mRNA | BC065318.1 | - | 6 - 7 | 539 | 65 | |
| BC153660.1 | - | 6 - 7 | 555 | 65 | ||
More Information
Set Membership: |
|

rab18a
Genomic Map