Assay Details
Gene Symbol: mcm7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212569.1 | NP_997734.1 | 948 | 88 | ||
GenBank mRNA | BC045497.1 | - | 948 | 88 | ||
BC065669.1 | - | 800 | 88 | |||
MH746781.1 | - | 868 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
