Assay Details
| Gene Symbol: ywhae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212605.2 | NP_997770.1 | 3 - 4 | 557 | 82 | |
| XM_021481333.1 | XP_021337008.1 | 3 - 4 | 514 | 82 | ||
| GenBank mRNA | BC045325.1 | - | 3 - 4 | 475 | 82 | |
| BC066763.1 | - | 3 - 4 | 458 | 82 | ||
| BC160627.1 | - | 3 - 4 | 462 | 82 | ||
| BC165123.1 | - | 3 - 4 | 408 | 82 | ||
More Information
Set Membership: |
|

ywhae1
Genomic Map