Assay Details
| Gene Symbol: rbb4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212610.1 | NP_997775.1 | 6 - 7 | 717 | 82 | |
| XM_009305881.3 | XP_009304156.1 | 5 - 6 | 968 | 82 | ||
| GenBank mRNA | BC052110.1 | - | 6 - 7 | 717 | 82 | |
| BC154778.1 | - | 6 - 7 | 733 | 82 | ||
More Information
Set Membership: |
|

rbb4l
Genomic Map