Assay Details
| Gene Symbol: ahsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212622.2 | NP_997787.2 | 1 - 2 | 311 | 112 | |
| GenBank mRNA | AY217758.2 | - | 1 - 2 | 240 | 112 | |
| BC085588.1 | - | 1 - 2 | 241 | 112 | ||
| BC165731.1 | - | 1 - 2 | 241 | 112 | ||
| EH504898.1 | - | 1 - 2 | 304 | 112 | ||
| EH520076.1 | - | 1 - 2 | 119 | 112 | ||
More Information
Set Membership: |
|

ahsg1
Genomic Map