Assay Details
| Gene Symbol: ahsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212622.2 | NP_997787.2 | 3 - 4 | 499 | 96 | |
| GenBank mRNA | AY217758.2 | - | 3 - 4 | 428 | 96 | |
| EH504898.1 | - | 3 - 4 | 492 | 96 | ||
More Information
Set Membership: |
|

ahsg1
Genomic Map