Assay Details
| Gene Symbol: ahsg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212622.2 | NP_997787.2 | 4 - 5 | 667 | 83 | |
| GenBank mRNA | AY217758.2 | - | 4 - 5 | 596 | 83 | |
| BC085588.1 | - | 4 - 5 | 597 | 83 | ||
| BC165731.1 | - | 4 - 5 | 597 | 83 | ||
| EH504898.1 | - | 4 - 5 | 660 | 83 | ||
| EH531076.1 | - | 3 - 4 | 915 | 83 | ||
More Information
Set Membership: |
|

ahsg1
Genomic Map