Assay Details
| Gene Symbol: pdap1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212632.2 | NP_997797.1 | 5 - 6 | 660 | 67 | |
| GenBank mRNA | BC052972.1 | - | 5 - 6 | 660 | 67 | |
| BC153633.1 | - | 5 - 6 | 456 | 67 | ||
| EH586204.1 | - | 5 - 6 | 656 | 67 | ||
More Information
Set Membership: |
|

pdap1a
Genomic Map