Assay Details
| Gene Symbol: lasp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212636.1 | NP_997801.1 | 5 - 6 | 531 | 56 | |
| GenBank mRNA | BC054945.1 | - | 5 - 6 | 462 | 56 | |
| BC066574.1 | - | 5 - 6 | 531 | 56 | ||
| BC155178.1 | - | 5 - 6 | 593 | 56 | ||
More Information
Set Membership: |
|

lasp1
Genomic Map