Assay Details
| Gene Symbol: aldh3a2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212649.1 | NP_997814.1 | 2 - 3 | 233 | 75 | |
| XM_009291620.3 | XP_009289895.1 | 2 - 3 | 295 | 75 | ||
| XM_017359216.2 | XP_017214705.1 | 2 - 3 | 251 | 75 | ||
| GenBank mRNA | AF254954.1 | - | 2 - 3 | 233 | 75 | |
| BC075877.1 | - | 2 - 3 | 222 | 75 | ||
| BC152640.1 | - | 2 - 3 | 240 | 75 | ||
| BC155171.1 | - | 2 - 3 | 251 | 75 | ||
More Information
Set Membership: |
|

aldh3a2a
Genomic Map