Assay Details
| Gene Symbol: csnk1e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212747.1 | NP_997912.1 | 8 - 9 | 1340 | 88 | |
| GenBank mRNA | BC065339.1 | - | 8 - 9 | 1340 | 88 | |
| BC155187.1 | - | 8 - 9 | 1344 | 88 | ||
| EU127824.1 | - | 7 - 8 | 1090 | 88 | ||
More Information
Set Membership: |
|

csnk1e
Genomic Map