Assay Details
| Gene Symbol: pou4f2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212807.1 | NP_997972.1 | 1 - 2 | 496 | 61 | |
| GenBank mRNA | AB122025.1 | - | 1 - 2 | 225 | 61 | |
| AY196176.1 | - | 1 - 2 | 496 | 61 | ||
| BC162312.1 | - | 1 - 2 | 492 | 61 | ||
| BC162336.1 | - | 1 - 2 | 492 | 61 | ||
More Information
Set Membership: |
|

pou4f2
Genomic Map