Assay Details
Gene Symbol: cnr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212820.2 | NP_997985.1 | 2 - 3 | 1134 | 97 | |
GenBank mRNA | AY148349.2 | - | 1 - 2 | 2067 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
