Assay Details
| Gene Symbol: glceb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212849.1 | NP_998014.1 | 2 - 3 | 730 | 69 | |
| XM_005168839.3 | XP_005168896.1 | 4 - 5 | 69 | |||
| GenBank mRNA | AY388517.1 | - | 2 - 3 | 730 | 69 | |
| BC163091.1 | - | 2 - 3 | 69 | |||
| BC163093.1 | - | 2 - 3 | 69 | |||
More Information
Set Membership: |
|

glceb
Genomic Map