Assay Details
Gene Symbol: rsrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212947.2 | NP_998112.2 | 3 - 4 | 366 | 68 | |
GenBank mRNA | BC067583.1 | - | 309 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
