Assay Details
| Gene Symbol: blzf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001144803.1 | NP_001138275.1 | 4 - 5 | 795 | 67 | |
| XM_021476915.1 | XP_021332590.1 | 5 - 6 | 832 | 67 | ||
| GenBank mRNA | BC055578.1 | - | 4 - 5 | 729 | 67 | |
| BC070020.1 | - | 4 - 5 | 739 | 67 | ||
More Information
Set Membership: |
|

blzf1
Genomic Map