Assay Details
| Gene Symbol: ppib | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213019.1 | NP_998184.1 | 4 - 5 | 689 | 77 | |
| GenBank mRNA | BC059560.1 | - | 4 - 5 | 556 | 77 | |
| BC071458.1 | - | 4 - 5 | 689 | 77 | ||
| BC154213.1 | - | 4 - 5 | 580 | 77 | ||
More Information
Set Membership: |
|

ppib
Genomic Map