Assay Details
| Gene Symbol: glrx5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213021.1 | NP_998186.1 | 1 - 2 | 348 | 61 | |
| GenBank mRNA | BC059659.1 | - | 1 - 2 | 348 | 61 | |
| DQ083329.1 | - | 1 - 2 | 299 | 61 | ||
More Information
Set Membership: |
|

glrx5
Genomic Map