Assay Details
| Gene Symbol: fgg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213054.1 | NP_998219.1 | 8 - 9 | 1151 | 62 | |
| GenBank mRNA | BC045868.1 | - | 8 - 9 | 1151 | 62 | |
| BC153662.1 | - | 8 - 9 | 1155 | 62 | ||
| BC165806.1 | - | 8 - 9 | 1149 | 62 | ||
More Information
Set Membership: |
|

fgg
Genomic Map