Assay Details
| Gene Symbol: eif3s6ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213128.1 | NP_998293.1 | 13 - 14 | 1651 | 115 | |
| GenBank mRNA | BC054626.1 | - | 13 - 14 | 1651 | 115 | |
More Information
Set Membership: |
|

eif3s6ip
Genomic Map