Assay Details
Gene Symbol: ap2s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213155.2 | NP_998320.1 | 2 - 3 | 313 | 65 | |
GenBank mRNA | BC058042.1 | - | 255 | 65 | ||
DT873506.1 | - | 2 - 3 | 317 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
