Assay Details
| Gene Symbol: vdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213246.1 | NP_998411.1 | 8 - 9 | 823 | 92 | |
| XM_005166743.4 | XP_005166800.1 | 9 - 10 | 914 | 92 | ||
| XM_005166744.4 | XP_005166801.1 | 8 - 9 | 911 | 92 | ||
| XM_005166745.2 | XP_005166802.1 | 10 - 11 | 951 | 92 | ||
| XM_005166746.3 | XP_005166803.1 | 9 - 10 | 948 | 92 | ||
| GenBank mRNA | BC065468.1 | - | 8 - 9 | 823 | 92 | |
| BC153608.1 | - | 8 - 9 | 793 | 92 | ||
More Information
Set Membership: |
|

vdac3
Genomic Map