Assay Details
Gene Symbol: sec13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213335.1 | NP_998500.1 | 8 - 9 | 948 | 70 | |
GenBank mRNA | BC054585.1 | - | 8 - 9 | 948 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
