Assay Details
| Gene Symbol: nudt9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213352.2 | NP_998517.2 | 5 - 6 | 687 | 82 | |
| GenBank mRNA | BC057417.1 | - | 5 - 6 | 638 | 82 | |
| BC152216.1 | - | 5 - 6 | 628 | 82 | ||
| EH449789.1 | - | 5 - 6 | 666 | 82 | ||
| EH523281.1 | - | 3 - 4 | 779 | 82 | ||
More Information
Set Membership: |
|

nudt9
Genomic Map