Assay Details
| Gene Symbol: nudt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213418.1 | NP_998583.1 | 3 - 4 | 292 | 101 | |
| GenBank mRNA | BC049486.1 | - | 3 - 4 | 292 | 101 | |
More Information
Set Membership: |
|

nudt1
Genomic Map