Assay Details
| Gene Symbol: ssbp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213450.1 | NP_998615.1 | 12 - 13 | 917 | 77 | |
| XM_005166809.4 | XP_005166866.1 | 13 - 14 | 1020 | 77 | ||
| XM_005166810.4 | XP_005166867.1 | 13 - 14 | 974 | 77 | ||
| XM_009303844.3 | XP_009302119.1 | 12 - 13 | 946 | 77 | ||
| GenBank mRNA | BC056827.1 | - | 12 - 13 | 917 | 77 | |
| GQ903695.1 | - | 13 - 14 | 890 | 77 | ||
More Information
Set Membership: |
|

ssbp3b
Genomic Map