Assay Details
| Gene Symbol: ssbp3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213450.1 | NP_998615.1 | 14 - 15 | 1014 | 85 | |
| GenBank mRNA | BC056827.1 | - | 14 - 15 | 1014 | 85 | |
| GQ903695.1 | - | 15 - 16 | 987 | 85 | ||
More Information
Set Membership: |
|

ssbp3b
Genomic Map