Assay Details
| Gene Symbol: eif3jb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_213502.1 | NP_998667.1 | 7 - 8 | 710 | 130 | |
| GenBank mRNA | BC044399.1 | - | 7 - 8 | 710 | 130 | |
| BC165515.1 | - | 7 - 8 | 692 | 130 | ||
More Information
Set Membership: |
|

eif3jb
Genomic Map