Assay Details
Gene Symbol: chmp2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213517.1 | NP_998682.1 | 5 - 6 | 684 | 66 | |
GenBank mRNA | BC045320.1 | - | 5 - 6 | 684 | 66 | |
BC171414.1 | - | 5 - 6 | 594 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
