Assay Details
| Gene Symbol: prrx1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214734.1 | NP_999899.1 | 3 - 4 | 723 | 78 | |
| GenBank mRNA | BC053228.1 | - | 3 - 4 | 723 | 78 | |
| BC165100.1 | - | 3 - 4 | 631 | 78 | ||
More Information
Set Membership: |
|

prrx1a
Genomic Map