Assay Details
| Gene Symbol: calm2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_214736.1 | NP_999901.1 | 5 - 6 | 467 | 72 | |
| GenBank mRNA | BC054600.1 | - | 5 - 6 | 467 | 72 | |
More Information
Set Membership: |
|

calm2b
Genomic Map