Assay Details
Gene Symbol: gpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214801.2 | NP_999966.2 | 4 - 5 | 845 | 85 | |
XM_009292455.3 | XP_009290730.1 | 4 - 5 | 834 | 85 | ||
GenBank mRNA | AY576999.1 | - | 4 - 5 | 827 | 85 | |
BC083538.1 | - | 4 - 5 | 831 | 85 | ||
BC150450.1 | - | 4 - 5 | 822 | 85 | ||
BC152270.1 | - | 4 - 5 | 793 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
