Assay Details
Gene Symbol: ttll11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077375.1 | NP_001070843.1 | 6 - 7 | 1615 | 141 | |
GenBank mRNA | BC124694.1 | - | 6 - 7 | 1615 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
