Assay Details
| Gene Symbol: ccl38a.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030173.1 | NP_001025344.1 | 1 - 2 | 95 | 132 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ccl38a.5| Gene Symbol: ccl38a.5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001030173.1 | NP_001025344.1 | 1 - 2 | 95 | 132 | |
Set Membership: |
|