Assay Details
Gene Symbol: opn1mw3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182892.2 | NP_878312.1 | 4 - 5 | 1015 | 119 | |
GenBank mRNA | BC075989.1 | - | 4 - 5 | 1015 | 119 | |
KT008396.1 | - | 4 - 5 | 940 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
