Assay Details
| Gene Symbol: hdhd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045267.2 | NP_001038732.2 | 2 | 1079 | 146 | |
| GenBank mRNA | BC116519.1 | - | 3 | 1254 | 146 | |
| BC133114.1 | - | 4 | 1097 | 146 | ||
More Information
Set Membership: |
|

hdhd3
Genomic Map