Assay Details
| Gene Symbol: eef2a.1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001045161.2 | NP_001038626.2 | 9 | 1647 | 126 | |
| GenBank mRNA | BC139576.1 | - | 11 | 1675 | 126 | |
More Information
Set Membership: |
|

eef2a.1
Genomic Map