Assay Details
| Gene Symbol: cdkn2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199994.1 | NP_001186923.1 | 2 - 3 | 771 | 101 | |
| GenBank mRNA | BC091928.1 | - | 2 - 3 | 777 | 101 | |
| BC128821.1 | - | 2 - 3 | 754 | 101 | ||
| BC160669.1 | - | 1 - 2 | 636 | 101 | ||
More Information
Set Membership: |
|

cdkn2c
Genomic Map