Assay Details
| Gene Symbol: NRSN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_023626282.1 | XP_023482050.1 | 738 | 62 | ||
| XM_023626283.1 | XP_023482051.1 | 653 | 62 | |||
| XM_023626284.1 | XP_023482052.1 | 817 | 62 | |||
| XM_023626285.1 | XP_023482053.1 | 255 | 62 | |||
More Information
Set Membership: |
|

NRSN2
Genomic Map