Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641295.1 | XP_023497063.1 | 341 | 74 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023641295.1 | XP_023497063.1 | 341 | 74 |