Assay Details
| Gene Symbol: HTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081782.2 | NP_001075251.1 | 1 | 1486 | 62 | |
| GenBank mRNA | AB264325.2 | - | 1 | 951 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HTR1A| Gene Symbol: HTR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081782.2 | NP_001075251.1 | 1 | 1486 | 62 | |
| GenBank mRNA | AB264325.2 | - | 1 | 951 | 62 | |
Set Membership: |
|