Assay Details
| Gene Symbol: TLR9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081790.2 | NP_001075259.1 | 1 - 2 | 101 | 63 | |
| GenBank mRNA | DQ390541.2 | - | 1 - 2 | 103 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081790.2 transcripts.
Set Membership: |
|

TLR9
Genomic Map