Assay Details
| Gene Symbol: SFTPA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081812.2 | NP_001075281.1 | 2 - 3 | 314 | 111 | |
| XM_014733440.1 | XP_014588926.1 | 553 | 111 | |||
| XM_014733441.1 | XP_014588927.1 | 473 | 111 | |||
| XM_023636212.1 | XP_023491980.1 | 375 | 111 | |||
| GenBank mRNA | AB015963.1 | - | 2 - 3 | 315 | 111 | |
More Information
Set Membership: |
|

SFTPA1
Genomic Map