Assay Details
Gene Symbol: AKR1C1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081908.1 | NP_001075377.1 | 1 - 2 | 107 | 85 | |
GenBank mRNA | AY955082.1 | - | 1 - 2 | 107 | 85 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081908.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
