Assay Details
Gene Symbol: LY49B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081923.1 | NP_001075392.1 | 1 - 2 | 43 | 60 | |
GenBank mRNA | AB120380.1 | - | 1 - 2 | 43 | 60 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001081923.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
