Assay Details
| Gene Symbol: CALCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081928.2 | NP_001075397.1 | 3 - 4 | 291 | 87 | |
| GenBank mRNA | AF257470.1 | - | 3 - 4 | 305 | 87 | |
More Information
Set Membership: |
|

CALCB
Genomic Map